Learn More
Selleck Chemical LLC Trastuzumab Emtansine(T-DM1)-1
SDP
Supplier:  Selleck Chemical LLC D40031MG
Encompass_Preferred
Trastuzumab Emtansine(T-DM1,Ado-Trastuzumab emtansine) is an antibody-drug conjugate (ADC) that incorporates the HER2-targeted antitumor properties of trastuzumab with the cytotoxic activity of the microtubule-inhibitory agent DM1.Trastuzumab emtansine has a molecular weight of 145kDa.This product is supplied as 10mg/ml PBS solution. This product is discontinued. We recommend the replacement products: D4003
Catalog No. 50-238-9757