Learn More
KRISHGEN BIOSYSTEMS Immunoassay for the estimation of Trastuzumab in serum and plasma. Validated against NIBSC standards. Can be used for generics and biosimilars PK studies.
Supplier:  KRISHGEN BIOSYSTEMS KBI1017
Encompass_Preferred
Pack Size: 1 x 96 wells. Assay Range: 10 - 640 ng/ml; Sensitivity: 8 ng/ml. Background: Trastuzumab is a humanized monoclonal antibody which targets Her-2/neu receptor. It binds to an extracellular domain of this receptor and inhibits HER2 homodimerization, thereby preventing HER2-mediated signaling. It may also upregulate cell cycle inhibitors such as p21 and p27. Trastuzumab is also thought to facilitate antibody-dependent cellular cytotoxicity, leading to the death of cells that express HER2. It is approved for use in treating breast cancers and is being tested for other cancers that overexpress HER2/neu. It is currently indicated for HER2 breast cancers and HER2 positive gastric cancer. This mAb-based Trastuzumab PK ELISA accurately quantifies trastuzumab in human samples. If you are looking to use animal samples, please contact us. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <10%.
Catalog No. NC2747086