Learn More
Cayman Chemical Traumatic Acid, Purity: ≥98%, CAS: 6402-36-4, 500 mg
SDP
Catalog No. 502896605
Encompass_Preferred
Traumatic acid is a product of the hydroperoxide lyase pathway in plants. It is a wound healing agent that stimulates cell division near a wound site to form a protective callus.{770}
Catalog No. 50-289-6605 Supplier Cayman Chemical Supplier No. 88820500 MG
Traumatic acid is a product of the hydroperoxide lyase pathway in plants. It is a wound healing agent that stimulates cell division near a wound site to form a protective callus.{770}