Learn More
Abcam Traut's reagent, RNA-protein crosslinking (sulfhydryl addition) agent, 1G
Catalog No. 502705107
Encompass_Preferred
MW 137.63 Da, Purity >90%. RNA-protein crosslinking (sulfhydryl addition) agent. Reacts with primary amines. Maintains charge properties similar to the original amino group.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5107 Supplier Abcam Supplier No. AB1456151G
MW 137.63 Da, Purity >90%. RNA-protein crosslinking (sulfhydryl addition) agent. Reacts with primary amines. Maintains charge properties similar to the original amino group.
The product is subject to the following: Abcam Restricted Use Statement