Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Travelscope
GREENER_CHOICESCI_ED

Catalog No. S89527 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89527 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89527 Supplier Supertek™ Supplier No. B10512S3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This 20x stereo Travelscope is a valuable tool for exploring anything from coins and stamps to the hidden world of nature. Because it is battery operated and light-weight it can go almost anywhere. A great field trip microscope. Discover more about the world in which we live.

  • 20x Stereo Microscope
  • Battery Powered
  • Requires 2 AA batteries (not included)
  • Light-weight
  • Size: 6.75 x 5.25 x 11 in.
  • Ages 7+

Specifications

Grade 1
For Use With (Equipment) AA Batteries
Product Type Supertek™ Travelscope
For Use With (Application) Field Trips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.