Learn More
Medchemexpress LLC HY-W018061 5mg Medchemexpress, Traxoprodil CAS:134234-12-1 Purity:>98%
SDP
Catalog No. 501934642
Encompass_Preferred
Medchemexpress, HY-W018061 5mg Traxoprodil CAS:134234-12-1 Traxoprodil (CP101,606) is a potent and selective NMDA antagonist and protect hippocampal neurons with an IC 50 of 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4642 Supplier Medchemexpress LLC Supplier No. HYW0180615MG
Medchemexpress, HY-W018061 5mg Traxoprodil CAS:134234-12-1 Traxoprodil (CP101,606) is a potent and selective NMDA antagonist and protect hippocampal neurons with an IC 50 of 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.