Learn More
Julabo USA Inc Each, tray and 4 test tube racks, 240 tubes 17mm Diameter. SW22/SW23.
SDP
Catalog No. 507534071
Encompass_Preferred
Bath Accessories, Julabo, Tray with 4 racks 8970380 for 240 tubes 100 x 16/17 mm, to +80°C. For use with shaking water baths: SW22 / SW23.
Catalog No. 50-753-4071 Supplier Julabo USA Inc Supplier No. 8960440
Bath Accessories, Julabo, Tray with 4 racks 8970380 for 240 tubes 100 x 16/17 mm, to +80°C. For use with shaking water baths: SW22 / SW23.