Learn More
Bellco Glass, Inc. Tray,30x30cm
SDP
Supplier:  Bellco Glass, Inc. 774001212
Encompass_Preferred
Standard tray with pegs, *NOT* a stacking tray. Trays fit 7740 and 7743 series rocker & 7777 series 12v rocker. The "peg and grommet" design permits rapid platform changing. . MADE IN THE USA. REPLACEMENT LINER: A502-193 SIZE: 30x30cm
Catalog No. 50-194-6106