Learn More
Julabo USA Inc Each, tray for 32 50mL Erlenmeyer flasks. SW22/SW23 shaking water baths.
SDP
Catalog No. 507534076
Encompass_Preferred
Bath Accessories, Julabo, Carrier trays for Erlenmeyer flasks, for 32 flasks, 50 mL. For use with shaking water bath models SW22, SW23.
Catalog No. 50-753-4076 Supplier Julabo USA Inc Supplier No. 8970361
Bath Accessories, Julabo, Carrier trays for Erlenmeyer flasks, for 32 flasks, 50 mL. For use with shaking water bath models SW22, SW23.