Learn More
Bellco Glass, Inc. Tray,51x51cm
SDP
Catalog No. 501946107
Encompass_Preferred
Standard tray with pegs, *NOT* a stacking tray. Trays fit 7740 and 7743 series rocker & 7777 series 12v rocker. The "peg and grommet" design permits rapid platform changing.. MADE IN THE USA SIZE: 51x51cm
Catalog No. 50-194-6107 Supplier Bellco Glass, Inc. Supplier No. 774002020
Standard tray with pegs, *NOT* a stacking tray. Trays fit 7740 and 7743 series rocker & 7777 series 12v rocker. The "peg and grommet" design permits rapid platform changing.. MADE IN THE USA SIZE: 51x51cm