Learn More
BURKLE INC Tray stainless steel, AISI 304, 18.11x11.81x0.79 in. (460x300x20mm)
Catalog No. 5000025981
Encompass_Preferred
Instruments, labware or samplers can be placed and transported on the sturdy stainless steel tray. The smooth surface is easy to clean.
The rectangular instrument tray is suitable for hygienic placing in the fields of chemistry, pharmaceuticals, cosmetics, animal feed and food & beverage.
Catalog No. 50-000-25981 Supplier BURKLE INC Supplier No. 42080003
Instruments, labware or samplers can be placed and transported on the sturdy stainless steel tray. The smooth surface is easy to clean.
The rectangular instrument tray is suitable for hygienic placing in the fields of chemistry, pharmaceuticals, cosmetics, animal feed and food & beverage.