Learn More
Bellco Glass, Inc. Tray Coversion Kit For Old Style Orb.Shakers
SDP
Catalog No. 501946161
Encompass_Preferred
This is a conversion kit for old style orbital shakers only. The old style design mounting plate was bolted to the platform tray. This kit will make the unit to a peg and grommet design. Only for 7744-Series Orbital Shakers. Contact Service/Repair for more information.. MADE IN THE USA
Catalog No. 50-194-6161 Supplier Bellco Glass, Inc. Supplier No. 774431020
This is a conversion kit for old style orbital shakers only. The old style design mounting plate was bolted to the platform tray. This kit will make the unit to a peg and grommet design. Only for 7744-Series Orbital Shakers. Contact Service/Repair for more information.. MADE IN THE USA