Learn More
Med Vet International Dynarex Dressing Change Tray, Sterile, 20/Case
Catalog No. 502831716
Encompass_Preferred
A convenient sterile kit for aseptic dressing changes. Components arranged in procedural sequence for convenience Cost-effective and disposable Not made with natural rubber latex Contents: (1)Instructions; (1)Non-Woven Face Mask; (1)Pair Vinyl Gloves with Wallet, Powder Free; (1)Polylined Drape, 13" × 18"; (1)Tape Measure, 36"; (3)Alcohol Swabsticks; (3)PVP Swabsticks; (1)Non-Woven Split Sponge, 4" × 4"; (1)Non-Adherent Sponge, 2" × 2"; (1)Gauze Sponge, 2" × 2"; (1)Gauze Sponge, 4" × 4"; (1)Roll Transparent Dressing Tape, ¾" × 18"; (1)Transparent Dressing, 4" × 4 ¾"; (1)Dressing Change Label
Catalog No. 50-283-1716 Supplier Med Vet International Supplier No. 4705
A convenient sterile kit for aseptic dressing changes. Components arranged in procedural sequence for convenience Cost-effective and disposable Not made with natural rubber latex Contents: (1)Instructions; (1)Non-Woven Face Mask; (1)Pair Vinyl Gloves with Wallet, Powder Free; (1)Polylined Drape, 13" × 18"; (1)Tape Measure, 36"; (3)Alcohol Swabsticks; (3)PVP Swabsticks; (1)Non-Woven Split Sponge, 4" × 4"; (1)Non-Adherent Sponge, 2" × 2"; (1)Gauze Sponge, 2" × 2"; (1)Gauze Sponge, 4" × 4"; (1)Roll Transparent Dressing Tape, ¾" × 18"; (1)Transparent Dressing, 4" × 4 ¾"; (1)Dressing Change Label