Learn More
Electron Microscopy Sciences Aluminum Embedding Tray for Polyethylene Molds
SDP
Catalog No. 5034083
Encompass_Preferred
Aluminum Embedding Tray is available with 6-position to hold these Individual Mold during polymerization. Comes with anodized blue and natural color.
Catalog No. 50-340-83 Supplier Electron Microscopy Sciences Supplier No. 7017804
Aluminum Embedding Tray is available with 6-position to hold these Individual Mold during polymerization. Comes with anodized blue and natural color.