Learn More
Fisherbrand™ Tray Frame for SterilElite Autoclave
Catalog No. STE2452019 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
SterilElite Autoclave STE16
SterilElite Autoclave STE24
SterilElite Autoclave STE40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
STE2452019 SterilElite Autoclave STE24
STE4052055 SterilElite Autoclave STE40
STE1652014 SterilElite Autoclave STE16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. STE2452019
Add to Cart
Add to Cart
A structural support system that holds multiple trays in a stable, organized configuration inside the autoclave chamber.
  • Enables multi-level loading (stacking trays vertically)
  • Ensures optimal steam circulation between trays
  • Provides stable positioning during sterilization cycles
  • Compatible with full tray sets (includes tray holders/spring holders)
  • Corrosion-resistant construction (stainless steel)
  • Stainless steel tray frame for efficiency and capacity optimization, with tray set used for uniform sterilization and organization
Specifications
Autoclavable Autoclavable
For Use With (Equipment) SterilElite Autoclave STE24
Product Type Tray Frame
Weight (English) 8 lb.
Features Enables Multi-level Loading (Stacking Trays Vertically)
Material Stainless Steel
Operating Pressure 134°C
Quantity 1 Ea.
Sterilization Time 20 min.