Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY INSERT MESH BSK F/MDL1200
Catalog No. NC3721512
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Mesh Baskets, For Model 1510; Capacity: 0.5 gal.; I.D.: 5.5 x 6 x 4 in., Capacity: 63.99 oz., 63.99 oz., 1.89L, Depth: 3 in., 7.62cm, Length: 5 in., 12.7cm, Width: 4.75 in., 12.06cm
Catalog No. NC3721512 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100916333
Add to Cart
Add to Cart
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Mesh Baskets, For Model 1510; Capacity: 0.5 gal.; I.D.: 5.5 x 6 x 4 in., Capacity: 63.99 oz., 63.99 oz., 1.89L, Depth: 3 in., 7.62cm, Length: 5 in., 12.7cm, Width: 4.75 in., 12.06cm