Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY INSERT MESH BSK F/MDL2800
Catalog No. NC3721514
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Mesh Baskets, For Model 2800; Capacity: 0.75 gal.; I.D.: 8 x 4 x 3 in., Capacity: 95.99 oz., 95.99 oz., 2.83L, Depth: 3 in., 7.62cm, Length: 8 in., 20.32cm, Width: 4 in., 10.16cm
Catalog No. NC3721514 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100916334
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Mesh Baskets, For Model 2800; Capacity: 0.75 gal.; I.D.: 8 x 4 x 3 in., Capacity: 95.99 oz., 95.99 oz., 2.83L, Depth: 3 in., 7.62cm, Length: 8 in., 20.32cm, Width: 4 in., 10.16cm