Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY INSERT MESH BSK F/MDL3200
Catalog No. NC3721516
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Mesh Baskets, For Model 3510; Capacity: 1.5 gal.; I.D.: 11.5 x 6 x 6 in., Capacity: 192 oz., 192 oz., 5.67L, Depth: 5 in., 12.7cm, Length: 10.5 in., 26.67cm, Width: 4.87 in., 12.36cm
Catalog No. NC3721516 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100916335
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Mesh Baskets, For Model 3510; Capacity: 1.5 gal.; I.D.: 11.5 x 6 x 6 in., Capacity: 192 oz., 192 oz., 5.67L, Depth: 5 in., 12.7cm, Length: 10.5 in., 26.67cm, Width: 4.87 in., 12.36cm