Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY INSERT PERFORT F/MDL 5200
Catalog No. NC3721557
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 5800; Capacity: 2.5 gal. (11.4L); I.D.: 11.5 x 9.5 x 6 in., Capacity: 11.4L (2.5 gal.), For Use With: Model 5800, Diameter Inner: 11.5 x 9.5 x 6 in., 29.2 x 24.1 x 15.2cm, Stainless Steel
Catalog No. NC3721557 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100410166
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 5800; Capacity: 2.5 gal. (11.4L); I.D.: 11.5 x 9.5 x 6 in., Capacity: 11.4L (2.5 gal.), For Use With: Model 5800, Diameter Inner: 11.5 x 9.5 x 6 in., 29.2 x 24.1 x 15.2cm, Stainless Steel