Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY INSERT SOLID MODEL 5200
Catalog No. NC3721553
Encompass_Preferred
Branson Ultrasonics Ultrasonic Cleaning Bath Accessory, Solid Insert Tray, For Use With: Model 5800 ultrasonic bath, Capacity: 2.5 gal., 9.4L, Depth: 6 in., 15.24cm, Dimensions: 10.5 x 8 in. (26.67 x 20.32cm), Includes: Built in handle, Length: 10.5 in., 26.67cm, Stainless Steel
Catalog No. NC3721553 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100410176
Branson Ultrasonics Ultrasonic Cleaning Bath Accessory, Solid Insert Tray, For Use With: Model 5800 ultrasonic bath, Capacity: 2.5 gal., 9.4L, Depth: 6 in., 15.24cm, Dimensions: 10.5 x 8 in. (26.67 x 20.32cm), Includes: Built in handle, Length: 10.5 in., 26.67cm, Stainless Steel