Learn More
Research Products International Corp Sterilizing Tray Cover, Fits 12 x 7 3/4 x 5 1/8 Inch Tray
SDP
Catalog No. 50212952
Encompass
For use with Sterilizing Tray 12 x 7 3/4 x 5 1/8 inches.
Polypropylene covers are 3.2 mm (1/8 inch) thick with a 25.4mm (1 inch) skirt that fits snugly over the rim of the tray. Trays and lids are steam autoclavable at 121°C (250°F).
Catalog No. 50-212-952 Supplier Research Products International Corp Supplier No. H16259
For use with Sterilizing Tray 12 x 7 3/4 x 5 1/8 inches.
Polypropylene covers are 3.2 mm (1/8 inch) thick with a 25.4mm (1 inch) skirt that fits snugly over the rim of the tray. Trays and lids are steam autoclavable at 121°C (250°F).