Learn More
Research Products International Corp Sterilizing Tray Cover, Fits 19 x 10 1/2 x 5 1/8 Inch Tray
SDP
Catalog No. 50212954
Encompass
For use with Sterilizing Tray 19 x 10 1/2 x 5 1/8 inches.
Polypropylene covers are 3.2 mm (1/8 inch) thick with a 25.4mm (1 inch) skirt that fits snugly over the rim of the tray. Trays and lids are steam autoclavable at 121ºC (250ºF).
Catalog No. 50-212-954 Supplier Research Products International Corp Supplier No. H16261
For use with Sterilizing Tray 19 x 10 1/2 x 5 1/8 inches.
Polypropylene covers are 3.2 mm (1/8 inch) thick with a 25.4mm (1 inch) skirt that fits snugly over the rim of the tray. Trays and lids are steam autoclavable at 121ºC (250ºF).