Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY PERF F/B1210 1/2GAL TANK
Catalog No. NC3721495
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 1800; Capacity: 0.5 gal. (2.27L); I.D.: 5.5 x 6 x 4 in., Capacity: 2.27L (0.5 gal.), For Use With: Model 1800, Diameter Inner: 5.5 x 6 x 4 in., 15 x 15.2 x 10.7cm, Stainless Steel
Catalog No. NC3721495 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100410160
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 1800; Capacity: 0.5 gal. (2.27L); I.D.: 5.5 x 6 x 4 in., Capacity: 2.27L (0.5 gal.), For Use With: Model 1800, Diameter Inner: 5.5 x 6 x 4 in., 15 x 15.2 x 10.7cm, Stainless Steel