Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY PERFO SS F/ULT CLN 5.5GL
Catalog No. NC3721503
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 8800; Capacity: 5.5 gal. (25L); I.D.: 17.5 x 9.5 x 6 in., Capacity: 25L (5.5 gal.), For Use With: Model 8800, Diameter Inner: 17.5 x 9.5 x 6 in., 44.45 x 24.1 x 15.2cm, Stainless Steel
Catalog No. NC3721503 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100410168
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 8800; Capacity: 5.5 gal. (25L); I.D.: 17.5 x 9.5 x 6 in., Capacity: 25L (5.5 gal.), For Use With: Model 8800, Diameter Inner: 17.5 x 9.5 x 6 in., 44.45 x 24.1 x 15.2cm, Stainless Steel