Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY PERFOR F/3210 1-1/2GAL TK
Catalog No. NC3721498
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 3800; Capacity: 1.5 gal. (6.8L); I.D.: 11.5 x 6 x 6 in., Capacity: 6.8L (1.5 gal.), For Use With: Model 3800, Diameter Inner: 9 x 5 x 3.75 in., 29.2 x 15.2 x 15.2cm, Stainless Steel
Catalog No. NC3721498 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100410164
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 3800; Capacity: 1.5 gal. (6.8L); I.D.: 11.5 x 6 x 6 in., Capacity: 6.8L (1.5 gal.), For Use With: Model 3800, Diameter Inner: 9 x 5 x 3.75 in., 29.2 x 15.2 x 15.2cm, Stainless Steel