Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY PERFOR F/B2210 3/4GAL TK
Catalog No. NC3721497
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 2800; Capacity: 0.75 gal. (3.4L); I.D.: 9.5 x 5.5 x 4 in., Capacity: 3.4L (0.75 gal.), For Use With: Model 2800, Stainless Steel
Catalog No. NC3721497 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100410162
Branson Ultrasonics Bransonic Ultrasonic Cleaning Bath Accessory, Perforated Trays, For use with Model 2800; Capacity: 0.75 gal. (3.4L); I.D.: 9.5 x 5.5 x 4 in., Capacity: 3.4L (0.75 gal.), For Use With: Model 2800, Stainless Steel