Learn More
Fisherbrand™ Tray Set for SterilElite
Catalog No. STE1652024 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
SterilElite Autoclave STE16
SterilElite Autoclave STE24
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
STE1652024 SterilElite Autoclave STE16
STE2452025 SterilElite Autoclave STE24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. STE1652024
Add to Cart
Add to Cart
A structural support system that holds multiple trays in a stable, organized configuration inside the autoclave chamber.
  • Enables multi-level loading (stacking trays vertically)
  • Ensures optimal steam circulation between trays
  • Provides stable positioning during sterilization cycles
  • Compatible with full tray sets (includes tray holders/spring holders)
  • Corrosion-resistant construction (stainless steel)
  • Stainless steel tray frame for efficiency and capacity optimization, with tray set used for uniform sterilization and organization
  • It includes 3 trays and a frame for the trays
Specifications
For Use With (Application) Autoclaving
For Use With (Equipment) SterilElite Autoclave STE16
Product Type Tray Set
Weight (English) 14 lb.
Material Stainless Steel
Quantity 1 Ea.
Sterilization Time 20 min.