Learn More
Electron Microscopy Sciences Fiberboard Tray Set With Plastic Boxes, 16 Box Set
SDP
Catalog No. 5033165
Encompass_Preferred
A compact, solid fiberboard Tray Box makes instituting a plastic box/sample storage system more convenient and efficient. To form a Tray Set, use plastic boxes for specimen storage, and then organize boxes within the fiberboard Tray Box. These sets eliminates misplacement or inadvertent sample loss, and allows for rapid retrieval of specimens. Includes catalog item 64309.
Holds: 16 Boxes
Catalog No. 50-331-65 Supplier Electron Microscopy Sciences Supplier No. 6435005
A compact, solid fiberboard Tray Box makes instituting a plastic box/sample storage system more convenient and efficient. To form a Tray Set, use plastic boxes for specimen storage, and then organize boxes within the fiberboard Tray Box. These sets eliminates misplacement or inadvertent sample loss, and allows for rapid retrieval of specimens. Includes catalog item 64309.
Holds: 16 Boxes