Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY SOLID SS F/ULTR CLN 1.5G
Catalog No. NC3721509
Encompass_Preferred
Branson Ultrasonics Ultrasonic Cleaning Bath Accessory, Solid Insert Tray, For Use With: Model 3800 ultrasonic bath, Capacity: 1.5 gal., 6.8L, Depth: 4 in., 10.16cm, Dimensions: 8.5 x 4.25 in. (21.59 x 10.79cm), Includes: Built in handle, Length: 8.5 in., 21.59cm, Stainless Steel
Catalog No. NC3721509 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100410174
Branson Ultrasonics Ultrasonic Cleaning Bath Accessory, Solid Insert Tray, For Use With: Model 3800 ultrasonic bath, Capacity: 1.5 gal., 6.8L, Depth: 4 in., 10.16cm, Dimensions: 8.5 x 4.25 in. (21.59 x 10.79cm), Includes: Built in handle, Length: 8.5 in., 21.59cm, Stainless Steel