Learn More
BRANSON ULTRASONICS CORPORATION (3P) TRAY SOLID SS F/ULTRASNC CLN G
Catalog No. NC3721505
Encompass_Preferred
Branson Ultrasonics Ultrasonic Cleaning Bath Accessory, Solid Insert Tray, For Use With: Model 1800 ultrasonic bath, Capacity: 0.5 gal., 2.7L, Depth: 3 in., 7.62cm, Dimensions: 4.25 x 4.25 in. (10.79 x 10.79cm), Includes: Built in handle, Length: 4.25 in., 10.79cm, Stainless Steel
Catalog No. NC3721505 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100410170
Branson Ultrasonics Ultrasonic Cleaning Bath Accessory, Solid Insert Tray, For Use With: Model 1800 ultrasonic bath, Capacity: 0.5 gal., 2.7L, Depth: 3 in., 7.62cm, Dimensions: 4.25 x 4.25 in. (10.79 x 10.79cm), Includes: Built in handle, Length: 4.25 in., 10.79cm, Stainless Steel