Description
- Perforated or slotted design for:
- Efficient steam penetration
- Uniform heat distribution
- Durable stainless steel construction
- Compatible with tray frames for stacked configurations
- Easy loading/unloading
Specifications
Specifications
| Autoclavable | Autoclavable |
| For Use With (Equipment) | SterilElite Autoclave STE40 |
| Product Type | Tray |
| Features | Venting Holes- Open During Sterilization To Ensure Full Steam Penetration Closed After Cycle To Maintain Sterility During Transport/Storage Equipped With Handles For Safe And Easy Carrying Protects Contents From Post-Sterilization Contamination |
| Quantity | 1 Ea. |
| Sterilization Time | 20 min. |