Learn More
Med Vet International Tracheostomy Care And Cleaning Trays, One Compartment, 20/Case, Dynd40610
Supplier:  Med Vet International DYND40610
Encompass_Preferred
Tracheostomy Clean & Care Trays: Each Medline tracheostomy clean and care tray features the latest components for efficient and convenient performance of this important infection control task. Designed by clinicians, these latex-free trays provide for the individual needs of each patient and caregiver. All of our trays also feature our super soft and absorbent Avant Gauze 4x4's and trach dressing. The peel-down Tyvek lid protects sterile contents during storage and handling.*Minimum Qualification for Medicare "Established Trach" Reimbursement This sterile set includes a triple compartment tray, one moisture proof drape, a pair of vinyl gloves, one 4" x 4", 6-ply premium trach dressing, five cotton-tip applicators, two 4" x 4", 4-ply premium gauze sponges, twill tape (1/2" x 34"), two pipe cleaners, and pliable cleaning brush. Basic Tracheostomy Clean & Care Trays.
Catalog No. 50-283-1556