Learn More
Med Vet International Pre-Connected Catheterization Trays
Catalog No. 502831688
Encompass_Preferred
Pre-Connected Catheterization Trays: These sterile kits are ideal for self-catheterization training or for when disposal of urine is not readily available. Includes water-soluble lubricating jelly, 1000cc collection bag, moisture-proof underpad, fenestrated drape, and specimen container with ID label. All CSR wrapped. Also Contains: 14Fr Vinyl Catheter, 5-pk PVP swabsticks, 2 Dry Swabsticks, Vinyl Gloves, Graduated Collection Basin.
Catalog No. 50-283-1688 Supplier Med Vet International Supplier No. DYND10400
Pre-Connected Catheterization Trays: These sterile kits are ideal for self-catheterization training or for when disposal of urine is not readily available. Includes water-soluble lubricating jelly, 1000cc collection bag, moisture-proof underpad, fenestrated drape, and specimen container with ID label. All CSR wrapped. Also Contains: 14Fr Vinyl Catheter, 5-pk PVP swabsticks, 2 Dry Swabsticks, Vinyl Gloves, Graduated Collection Basin.