Learn More
Cole-Parmer Cole-Parmer Stainless steel utility tray, 19-1/8"L x 12-5/8"W
Catalog No. 50200180 Shop All Cole-Parmer Products
Encompass_Preferred
Stainless steel utility trays serve multiple purposes in the laboratory. These heavy-gauge 304 stainless steel trays can withstand temperatures up to 650°F (343°C). Keep several on hand to transport supplies, instruments, and other items.
Catalog No. 50-200-180 Supplier Cole-Parmer Supplier No. 0727719
Stainless steel utility trays serve multiple purposes in the laboratory. These heavy-gauge 304 stainless steel trays can withstand temperatures up to 650°F (343°C). Keep several on hand to transport supplies, instruments, and other items.