Learn More
Kuhner Shaker Inc TRAY WITH FIXED CLAMPS E-100ML
Catalog No. 502570268
Encompass
Tray with fixed clamps E-100mL; holds 39 flasks
Catalog No. 50-257-0268 Supplier Kuhner Shaker Inc Supplier No. SM320100
Tray with fixed clamps E-100mL; holds 39 flasks