Learn More
Kuhner Shaker Inc TRAY WITH FIXED CLAMPS E-200ML
Catalog No. 502570265
Encompass
Tray with fixed clamps E-200mL; Holds 20 flasks
Catalog No. 50-257-0265 Supplier Kuhner Shaker Inc Supplier No. SM320200
Tray with fixed clamps E-200mL; Holds 20 flasks