Learn More
Kuhner Shaker Inc TRAY WITH FIXED CLAMPS EX-1500
Catalog No. 502569992
Encompass
Tray with fixed clamps EX-1500; holds 8 flasks
Catalog No. 50-256-9992 Supplier Kuhner Shaker Inc Supplier No. SMX321500
Tray with fixed clamps EX-1500; holds 8 flasks