Learn More
Kuhner Shaker Inc TRAY WITH FIXED CLAMPS EX-6000
Catalog No. 502569987
Encompass
Tray with fixed clamps EX-6000; holds 2 flasks
Catalog No. 50-256-9987 Supplier Kuhner Shaker Inc Supplier No. SMX326000
Tray with fixed clamps EX-6000; holds 2 flasks