Learn More
Kuhner Shaker Inc TRAY WITH FIXED CLAMPS F-100ML
Catalog No. 502570252
Encompass
Tray with fixed clamps F-100mL; holds 74 flasks
Catalog No. 50-257-0252 Supplier Kuhner Shaker Inc Supplier No. SM330100
Tray with fixed clamps F-100mL; holds 74 flasks