Learn More
Kuhner Shaker Inc TRAY WITH FIXED CLAMPS F-50ML
Catalog No. 502570253
Encompass
Tray with fixed clamps F-50mL; holds 100 flasks
Catalog No. 50-257-0253 Supplier Kuhner Shaker Inc Supplier No. SM330050
Add to Cart
Add to Cart
Request a Quote
Tray with fixed clamps F-50mL; holds 100 flasks