Learn More
Kuhner Shaker Inc TRAY WITH PINS F-V5000/7
Catalog No. NC1940147
Encompass
Tray with Pins F-V5000/7..F Tray with pins to hold seven flasks ..(3L Corning, 5L Corning, 5L Thompson)..Max speed 150RPM at 50mm 200 at 25mm
Catalog No. NC1940147 Supplier Kuhner Shaker Inc Supplier No. SM335000S
Tray with Pins F-V5000/7..F Tray with pins to hold seven flasks ..(3L Corning, 5L Corning, 5L Thompson)..Max speed 150RPM at 50mm 200 at 25mm