Learn More
Medchemexpress LLC Trazodone-d6 (hydrochloride) | 1181578-71-1 | 99.6% | 1 MG
SDP
Catalog No. 5000407939
Encompass_Preferred
Trazodone-d6 (hydrochloride) is the deuterium labeled Trazodone hydrochloride.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07939 Supplier Medchemexpress LLC Supplier No. HYB0478AS1MG
Trazodone-d6 (hydrochloride) is the deuterium labeled Trazodone hydrochloride.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS