Learn More
Medchemexpress LLC Trazodone-d6 hydrochloride | 1181578-71-1 | MFCD09841350 | >99.6% | 5 MG
SDP
Catalog No. 5000407097
Encompass_Preferred
Trazodone-d6 hydrochloride is a deuterium-labeled form of Trazodone hydrochloride. This compound can be utilized as a tracer and an internal standard for quantitative analysis through methods like Nuclear Magnetic Resonance (NMR), Gas Chromatography-Mass Spectrometry (GC-MS), or Liquid Chromatography-Mass Spectrometry (LC-MS). Deuteration, which involves incorporating stable heavy isotopes such as deuterium into drug molecules, is known for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Functions as a tracer and internal standard
  • Applicable in quantitative analysis using NMR, GC-MS, or LC-MS
  • Impacts pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07097 Supplier Medchemexpress LLC Supplier No. HYB0478AS5MG
Trazodone-d6 hydrochloride is a deuterium-labeled form of Trazodone hydrochloride. This compound can be utilized as a tracer and an internal standard for quantitative analysis through methods like Nuclear Magnetic Resonance (NMR), Gas Chromatography-Mass Spectrometry (GC-MS), or Liquid Chromatography-Mass Spectrometry (LC-MS). Deuteration, which involves incorporating stable heavy isotopes such as deuterium into drug molecules, is known for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Functions as a tracer and internal standard
  • Applicable in quantitative analysis using NMR, GC-MS, or LC-MS
  • Impacts pharmacokinetic and metabolic profiles of drugs