Learn More
Selleck Chemical LLC Trazodone HCl 50mg 25332-39-2 AF-1161, KB-831
SDP
Catalog No. 501362589
Encompass_Preferred
Trazodone HCl (AF-1161, KB-831) is an antidepressant belonging to the class of serotonin receptor antagonists and reuptake inhibitors for treatment of anxiety disorders. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2589 Supplier Selleck Chemical LLC Supplier No. S258250MG
Trazodone HCl (AF-1161, KB-831) is an antidepressant belonging to the class of serotonin receptor antagonists and reuptake inhibitors for treatment of anxiety disorders. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.