Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trazodone hydorchloride, 99.9%, MP Biomedicals™

Catalog No. ICN19151805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19151805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19151805 Supplier MP Biomedicals, Inc Supplier No. 0219151805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antidepressant that acts as a serotonin uptake inhibitor

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25332-39-2
Molecular Formula C19H23Cl2N5O
Molecular Weight (g/mol) 408.327
InChI Key OHHDIOKRWWOXMT-UHFFFAOYSA-N
Synonym trazodone hydrochloride, trazodone hcl, desyrel, bimaran, molipaxin, devidon, pragmazone, thombran, tombran, tritico
PubChem CID 62935
ChEBI CHEBI:9655
IUPAC Name 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
SMILES C1CN(CCN1CCCN2C(=O)N3C=CC=CC3=N2)C4=CC(=CC=C4)Cl.Cl

Specifications

Color White
Assay Percent Range 0.999
Quantity 5 g
Formula Weight 408.3
Percent Purity 99.9%
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Trazodone hydorchloride
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.