Learn More
Medchemexpress LLC Trazodone hydrochloride | 25332-39-2 | 10 G
SDP
Catalog No. 5000405263
Encompass_Preferred
Trazodone hydrochloride (AF-1161) is a triazolopyridine derivative that belongs to the class of serotonin receptor antagonists and reuptake inhibitors (SARIs). It has antidepressant and anti-insomnious activity and exerts antagonistic properties against α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects.
  • Serotonin receptor antagonist and reuptake inhibitor
  • Exhibits antidepressant and anti-insomnious activity
  • Antagonistic properties against α1- and α2-adrenergic receptors and histamine H1 receptors
  • Minimal anticholinergic effects
Catalog No. 50-004-05263 Supplier Medchemexpress LLC Supplier No. HYB047810G
Trazodone hydrochloride (AF-1161) is a triazolopyridine derivative that belongs to the class of serotonin receptor antagonists and reuptake inhibitors (SARIs). It has antidepressant and anti-insomnious activity and exerts antagonistic properties against α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects.
  • Serotonin receptor antagonist and reuptake inhibitor
  • Exhibits antidepressant and anti-insomnious activity
  • Antagonistic properties against α1- and α2-adrenergic receptors and histamine H1 receptors
  • Minimal anticholinergic effects