Learn More
Medchemexpress LLC Trazodone (hydrochloride) | 25332-39-2 | 99.6% | 100 MG
SDP
Catalog No. 5000405525
Encompass_Preferred
Trazodone (hydrochloride) is a triazolopyridine derivative that acts as a serotonin receptor antagonist and reuptake inhibitor (SARI). It exhibits antidepressant and anti-insomnia activity, alongside antagonistic properties against α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects. This product is intended for use as a laboratory chemical and for substance manufacturing, and is stable under recommended storage conditions.
  • Triazolopyridine derivative
  • Serotonin receptor antagonist and reuptake inhibitor (SARI)
  • Demonstrates antidepressant and anti-insomnia activity
  • Exhibits antagonistic properties against α1- and α2-adrenergic and histamine H1 receptors
  • Minimal anticholinergic effects
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-004-05525 Supplier Medchemexpress LLC Supplier No. HYB0478R100MG
Trazodone (hydrochloride) is a triazolopyridine derivative that acts as a serotonin receptor antagonist and reuptake inhibitor (SARI). It exhibits antidepressant and anti-insomnia activity, alongside antagonistic properties against α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects. This product is intended for use as a laboratory chemical and for substance manufacturing, and is stable under recommended storage conditions.
  • Triazolopyridine derivative
  • Serotonin receptor antagonist and reuptake inhibitor (SARI)
  • Demonstrates antidepressant and anti-insomnia activity
  • Exhibits antagonistic properties against α1- and α2-adrenergic and histamine H1 receptors
  • Minimal anticholinergic effects
  • Stable under recommended storage conditions
  • For research use only