Learn More
Medchemexpress LLC Trazodone hydrochloride | 25332-39-2 | 99.6% | 5 G
SDP
Catalog No. 5000409425
Encompass_Preferred
Trazodone hydrochloride is a triazolopyridine derivative that acts as a serotonin receptor antagonist and reuptake inhibitor (SARI). It demonstrates anti-depressant and anti-insomnious activity by antagonizing α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects. It is suitable for various laboratory applications and research.
  • Exhibits anti-depressant and anti-insomnious activity
  • Acts as an antagonist against α1- and α2-adrenergic receptors
  • Antagonizes histamine H1 receptors
  • Demonstrates minimal anticholinergic effects
  • Binds to 5-HT1A receptor
  • Reduces expression of neuroinflammatory markers
  • Increases mRNA expression of BDNF and CREB
Catalog No. 50-004-09425 Supplier Medchemexpress LLC Supplier No. HYB04785G
Trazodone hydrochloride is a triazolopyridine derivative that acts as a serotonin receptor antagonist and reuptake inhibitor (SARI). It demonstrates anti-depressant and anti-insomnious activity by antagonizing α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects. It is suitable for various laboratory applications and research.
  • Exhibits anti-depressant and anti-insomnious activity
  • Acts as an antagonist against α1- and α2-adrenergic receptors
  • Antagonizes histamine H1 receptors
  • Demonstrates minimal anticholinergic effects
  • Binds to 5-HT1A receptor
  • Reduces expression of neuroinflammatory markers
  • Increases mRNA expression of BDNF and CREB