Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trazodone (hydrochloride) | 25332-39-2 | 99.6% | 50 MG
SDP

Catalog No. 5000405526
Encompass_Preferred

This product is a triazolopyridine derivative, classified as a serotonin receptor antagonist and reuptake inhibitor (SARI). It demonstrates antidepressant and anti-insomnia activities by antagonizing α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects. It is intended for research use only.

  • Classified as a serotonin receptor antagonist and reuptake inhibitor (SARI)
  • Exhibits antidepressant and anti-insomnia activity
  • Antagonizes α1- and α2-adrenergic receptors
  • Antagonizes histamine H1 receptors
  • Minimal anticholinergic effects
  • Purity of 99.60%
  • Molecular formula: C19H23Cl2N5O
  • Appears as a white to off-white solid
  • Soluble in DMSO and water
  • Binds to 5-HT1A receptor with high affinity
  • Inhibits key signaling pathways for neuroinflammatory markers
  • Increases BDNF and CREB expression

Catalog No. 50-004-05526 Supplier Medchemexpress LLC Supplier No. HYB0478R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a triazolopyridine derivative, classified as a serotonin receptor antagonist and reuptake inhibitor (SARI). It demonstrates antidepressant and anti-insomnia activities by antagonizing α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects. It is intended for research use only.

  • Classified as a serotonin receptor antagonist and reuptake inhibitor (SARI)
  • Exhibits antidepressant and anti-insomnia activity
  • Antagonizes α1- and α2-adrenergic receptors
  • Antagonizes histamine H1 receptors
  • Minimal anticholinergic effects
  • Purity of 99.60%
  • Molecular formula: C19H23Cl2N5O
  • Appears as a white to off-white solid
  • Soluble in DMSO and water
  • Binds to 5-HT1A receptor with high affinity
  • Inhibits key signaling pathways for neuroinflammatory markers
  • Increases BDNF and CREB expression

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.