Learn More
Medchemexpress LLC Trazodone hydrochloride | 25332-39-2 | 99.6% | 500 MG
SDP
Catalog No. 5000407942
Encompass_Preferred
Trazodone hydrochloride is a triazolopyridine derivative classified as a serotonin receptor antagonist and reuptake inhibitor (SARI). It demonstrates anti-depressant and anti-insomnious activity. The compound exerts antagonistic properties against α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects.
  • Serotonin receptor antagonist and reuptake inhibitor (SARI)
  • Exhibits anti-depressant and anti-insomnious activities
  • Antagonizes α1- and α2-adrenergic receptors and histamine H1 receptors
  • Minimal anticholinergic effects
  • Binds to 5-HT1A receptor
  • Reduces expression of neuroinflammatory markers
  • Increases mRNA expression of BDNF and CREB
  • Demonstrates neuroprotective effects
Catalog No. 50-004-07942 Supplier Medchemexpress LLC Supplier No. HYB0478500MG
Trazodone hydrochloride is a triazolopyridine derivative classified as a serotonin receptor antagonist and reuptake inhibitor (SARI). It demonstrates anti-depressant and anti-insomnious activity. The compound exerts antagonistic properties against α1- and α2-adrenergic receptors and histamine H1 receptors, with minimal anticholinergic effects.
  • Serotonin receptor antagonist and reuptake inhibitor (SARI)
  • Exhibits anti-depressant and anti-insomnious activities
  • Antagonizes α1- and α2-adrenergic receptors and histamine H1 receptors
  • Minimal anticholinergic effects
  • Binds to 5-HT1A receptor
  • Reduces expression of neuroinflammatory markers
  • Increases mRNA expression of BDNF and CREB
  • Demonstrates neuroprotective effects